Alkaline running buffer solution

Supplier: Thermo Fisher Scientific

J63170.AP
J63170.APEA 60.2 EUR
J63170.AP
Alkaline running buffer solution
Buffers Electrophoresis Buffers Running Buffers

Physical Form: Liquid
UN No.:UN1824


Contains 0.5 M NaOH and 50 mM EDTA

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR