Sodium glutamate

Supplier: Thermo Fisher Scientific

J63424.09 J63424.14
J63424.09EA 49.7 EUR
J63424.09 J63424.14
Sodium glutamate
Sodium glutamate

CAS No.: 142-47-2 Molecular Formula: C5H8NNaO4 Formula Weight: 169.11 Physical Form: Powder Appearance: White to off white MDL No.: MFCD00013074

Formula: C₅H₈NNaO₄
Storage Temperature: Ambient
MDL Number: MFCD00013074
CAS Number: 142-47-2
EINECS: 205-538-1
Merck Index: 14,6254

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR