AMP, 0.2 M Buffer Solutions

Supplier: Thermo Fisher Scientific

J63988.AEEA 54.4 EUR
J63988.AE
AMP, 0.2 M Buffer Solutions
Buffers Biological Buffers

2-Amino-2-methyl-1-propanol buffer, colorless and odorless.


CAS No.: 124-68-5
MDL No.: MFCD00008051


Caution: Storage Temperature: 2 to 8 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR