5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%

Supplier: Thermo Fisher Scientific

J64451.MD J64451.03
J64451.MDEA 135 EUR
J64451.MD J64451.03
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Formula: C₈H₄BrClNNa₂O₄P
Storage Temperature: Freezer
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR