Netilmicin sulphate 595 µg/mg

Supplier: Thermo Fisher Scientific
Danger

J66302.MA J66302.MB
J66302.MAEA 166 EUR
J66302.MA J66302.MB
Netilmicin sulphate 595 µg/mg
Netilmicin sulphate

Netilmicin sulphate is used as a broad spectrum antibiotic active against most gram-negative and some gram-positive bacteria.


  • Light Sensitive
  • Hygroscopic


It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studies.


White to off-white powder


Soluble in water


Caution: For research use only. Not intended for animal or human diagnostic or therapeutic use.

Formula: C₂₁H₄₁N₅O₇·⁵/₂H₂SO₄
Boiling Pt: 868 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01311715
CAS Number: 56391-57-2
EINECS: 260-147-3
Merck Index: 14,6479

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR