Gibberellic acid 90% A4+A7

Supplier: Thermo Fisher Scientific

Synonyms: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid

J66661.03 J66661.06
J66661.03EA 148 EUR
J66661.03 J66661.06
Gibberellic acid 90% A4+A7
Gibberellic acid

Purity: 90%
Storage Temperature: −30 to −10 °C
Flash Point: >110 °C
Physical Form: Powder
Boiling Point: 582°
Solubility: Soluble in methanol, ethanol, or acetone. Also soluble in dilute solutions of sodium bicarbonate or sodium acetate

Formula: C₁₉H₂₂O₆
MW: 346.38 g/mol
Boiling Pt: 582 °C
Melting Pt: 234
Flash Pt: >110 °C(230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00079329
CAS Number: 77-06-5
Merck Index: 13,04430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR