Escin 98

Supplier: Thermo Fisher Scientific
Warning

J66968.03 J66968.06 J66968.09
J66968.03EA 98.1 EUR
J66968.03 J66968.06 J66968.09
Escin 98
Escin
Formula: C₅₅H₈₆O₂₄
MW: 1131.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076054
CAS Number: 6805-41-0
EINECS: 229-880-6
Merck Index: 14,3696

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR