TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+% for molecular biology

Supplier: Thermo Fisher Scientific

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

J67233.22
J67233.22EA 0 EUR
J67233.22
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 99+% for molecular biology
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)

Applications: For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.


Applications: For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Melting Pt: ∼ 152 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR