Sodium dodecyl sulphate (SDS) solution 10%, Ultrapure

Supplier: Thermo Fisher Scientific

J77504.K2 J77504.AE J77504.AP
J77504.K2EA 172 EUR
J77504.K2 J77504.AE J77504.AP
Sodium dodecyl sulphate (SDS) solution 10%, Ultrapure
Sodium dodecyl sulphate (SDS)

10% (w/v) solution of sodium dodecyl sulfate (SDS) in high purity dH2O. Solution is 0.2μm filtered and dispensed into durable, square Nalgene bottles.
A(280 nm, 3%, 1 cm): ≤ 0.1
A(230 nm, 3%, 1 cm): ≤ 0.2


Caution: Note: Precipitation of SDS is not unusual. Warm gently to redissolve.

Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR