Cole-Parmer® WB-400 Series Digital General-Purpose Water Bath
Supplier: Cole-Parmer
12105-87
JENW12105-87EA 1760 EUR
JENW12105-87
Cole-Parmer® WB-400 Series Digital General-Purpose Water Bath
Baths Water Baths, Unstirred
Get digital accuracy at an analogue price.
  • Recall settings at the touch of a button - program up to 5 commonly used set points
  • Know when process is complete with built-in timer
  • Easy-to-read TFT display shows choice of °C or °F
  • Features high-temperature alarm and safety shutdown
Cole-Parmer utility baths combine precise temperature control and excellent temperature stability to provide reliable heating. These baths feature dual thermostats - a primary microprocessor-based temperature controller that provides ±0,1 °C stability (±0,2 °C uniformity), andMore Product Information