Jenway® pH Electrodes
Supplier: Cole-Parmer
99960-41 68006-40 59530-70 68006-37 99960-34 10090-75 99960-39 99960-44 90269-17
924049.EA 246 EUR
924049. JENW68006-40 JENW59530-70 JENW68006-37 JENW99960-34 JENW10090-75 JENW99960-39 JENW99960-44 JENW90269-17
Jenway® pH Electrodes
Electrodes pH Electrodes
A variety of pH electrodes for your Jewnay meters.
  • Choose the right electrode for your application
These electrodes are designed for standard measurements of general solutions. The epoxy body makes it especially suitable for field measurements or use in education.