Caps
Supplier: VICI JOUR RESEARCH
JR-S-11001 JR-S-11101 JR-S-11103 JR-S-11201 JR-S-11202 JR-S-11203 JR-S-11102 JR-S-11002 JR-S-11003 JR-S-15003 JR-S-15002 JR-S-15001 JR-S-13003 JR-S-13002 JR-S-13001 JR-S-11006 JR-S-11005 JR-S-14002
JOURJR-S-11001EA 89.8 EUR
JOURJR-S-11001 JOURJR-S-11101 JOURJR-S-11103 JOURJR-S-11201 JOURJR-S-11202 JOURJR-S-11203 JOURJR-S-11102 JOURJR-S-11002 JOURJR-S-11003 JOURJR-S-15003 JOURJR-S-15002 JOURJR-S-15001 JOURJR-S-13003 JOURJR-S-13002 JOURJR-S-13001 JOURJR-S-11006 JOURJR-S-11005 JOURJR-S-14002
Caps
Closures Safety Caps and Safety Waste Caps
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable