Polycarbonate retractable eyeshield, EVOSpec®
Supplier: JSP
ANT010-200-000
JSPAANT010-200-000EA 0 EUR
JSPAANT010-200-000
Polycarbonate retractable eyeshield, EVOSpec®
Head Protection Safety Helmets
Easy assembly kit for adding a clear, polycarbonate, retractable eyeshield to existing Evolution® helmets.
  • Polycarbonate retractable eye shield
  • 6 Base curve lens offers optimum coverage for all users
  • Low profile bi-polymer nose clip for improved comfort and security
  • Factory fit and/or user replaceable
  • Polypropylene arms
  • Anti-fog and anti-scratch treatment
Stylish design, retracts neatly and securely into helmet when not in use. Compatible with Evolution® safety helmets (EVOLite® / EVO3®/EVO5®).
Certifications: Conforms to EN 166.1.F.