VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline), USP sterile
Supplier: VWR Chemicals
K813-500MLEA 120 EUR
K813-500ML
VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline), USP sterile
Buffers Cell Culture Buffers
  • No DNases, RNases or proteases detected
  • Sterile
Product tested according to USP specification
pH @ 25 °C (1:10 water)    6,6 to 7,2