Antibacterial hand cleansers, KLEENEX®
Supplier: KIMBERLY CLARK
6336
KIMB6336EA 203 EUR
KIMB6336
Antibacterial hand cleansers, KLEENEX®
Skincare Soaps and Skin Cleansers
This antibacterial liquid hand cleanser is effective against a range of micro-organisms.
  • Kind to skin when hands are washed frequently
  • Reduces the risk of contamination, infection and illness
  • Dispenses the ideal amount of soap, prevents from overuse
  • Triclosan- , dye- and fragrance-free liquid gel
  • In strong, hygienically sealed 1-litre cassettes
  • Cassettes give up to 2500 shots of soap and are recyclable (when pump is removed)
It is ideal for washrooms in hygiene-critical areas such as healthcare, food processing and food service environments.