Culture tubes
Supplier: KLEINFELD
73500-18150 1217639
KLEI73500-18150EA 168 EUR
KLEI73500-18150 KLEI1217639
Culture tubes
Tubes Culture Tubes
Manufactured from 51 expansion borosilicate glass.
  • 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • The 6×50 mm size is often referred to as a 'Durham tube'
These premium quality tubes feature sturdy, uniform bottoms and consistent lengths.More Product Information