Bench mounted gas taps
Supplier: KOETTERMANN
733-00053 733-00054 733-00055 733-00056 733-00057 733-00058 733-00059 733-00060 733-00061
KOTT733-00053EA 0 EUR
KOTT733-00053 KOTT733-00054 KOTT733-00055 KOTT733-00056 KOTT733-00057 KOTT733-00058 KOTT733-00059 KOTT733-00060 KOTT733-00061
Bench mounted gas taps
Faucets and Sinks Faucets
These bench mounted gas taps are sealed by EPDM (ethylene propylene diene terpolymer) which is an extremely durable synthetic rubber roofing membrane.
Order Now
SPECIFICATIONS
Environmentally Preferable