5-Chloro-1-pentyne ≥98%

Supplier: Thermo Fisher Scientific
Danger

L02835.06 L02835.14
L02835.06EA 70.3 EUR
L02835.06 L02835.14
5-Chloro-1-pentyne ≥98%
5-Chloro-1-pentyne

Purity: 98% CAS No.: 14267-92-6 Molecular Formula: C5H7Cl Formula Weight: 102.56 Storage Temperature: 2°C to 8°C Flash Point: 15°(59°F) Refractive Index: 1.4440 Odour: Recognizable Boiling Point: 113° Beilstein Reference: 1736710 UN No.: 1993 MDL No.: MFCD00001014

Formula: C₅H₇Cl
MW: 102.56 g/mol
Boiling Pt: 113 °C
Density: 0.968
Flash Pt: 15 °C (59 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001014
CAS Number: 14267-92-6
EINECS: 238-159-5
UN: 1993

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR