Methylsulphonylacetonitrile ≥98%

Supplier: Thermo Fisher Scientific
Warning

L03280.09 L03280.18
L03280.09EA 85.5 EUR
L03280.09 L03280.18
Methylsulphonylacetonitrile ≥98%
Methylsulphonylacetonitrile

Purity: 98+% CAS No.: 2274-42-2 Molecular Formula: C3H5NO2S Formula Weight: 119.14 Melting Point: 82-86° Beilstein Reference: 969929 MDL No.: MFCD00014743

Formula: C₃H₅NO₂S
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00014743
CAS Number: 2274-42-2
EINECS: 218-888-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR