m-Toluenesulphonic acid monohydrate 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Methylbenzenesulphonic acid hydrate

L03419.06 L03419.14
L03419.06EA 116 EUR
L03419.06 L03419.14
m-Toluenesulphonic acid monohydrate 97%
m-Toluenesulphonic acid monohydrate

Purity: 97% CAS No.: 312619-56-0 Molecular Formula: C7H8O3S.H2O Formula Weight: 190.22 (172.20anhy) Refractive Index: 1.5270 Boiling Point: 165°/0.1mm Beilstein Reference: 2208421 UN No.: UN2586 MDL No.: MFCD00167130

Formula: C₇H₈O₃S·H₂O
MW: 190.22 g/mol (172.20 anhy)
Boiling Pt: 165 °C (0.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00167130
CAS Number: 312619-56-0
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR