m-Xylene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,3-Dimethylbenzene

L03788.AE L03788.AP L03788.0F
L03788.AEEA 30.6 EUR
L03788.AE L03788.AP L03788.0F
m-Xylene ≥99%
m-Xylene

Purity: 99% CAS No.: 108-38-3 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 25°(77°F) Refractive Index: 1.4970 Odour: Mild Melting Point: -47° Boiling Point: 138-139° Beilstein Reference: 605441 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00008536

Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 138…139 °C
Melting Pt: –47 °C
Density: 0.866
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008536
CAS Number: 108-38-3
EINECS: 203-576-3
UN: 1307
ADR: 3 II
Merck Index: 12,10214

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR