2-Ethylhexyl acrylate ≥98% stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Acrylic acid 2-ethylhexyl ester

L03854.AE L03854.AP L03854.0F
L03854.AEEA 28.9 EUR
L03854.AE L03854.AP L03854.0F
2-Ethylhexyl acrylate ≥98% stabilised
2-Ethylhexyl acrylate

Purity: 98% CAS No.: 103-11-7 Molecular Formula: C11H20O2 Formula Weight: 184.28 Flash Point: 79°(174°F) Refractive Index: 1.4360 Melting Point: -90° Boiling Point: 130°/50mm Beilstein Reference: 1765828 MDL No.: MFCD00009495

Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 130 °C (50 mmHg)
Melting Pt: –90 °C
Density: 0.885
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009495
CAS Number: 103-11-7
EINECS: 203-080-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR