1,2-Bis(dimethylamino)methane 99%

Supplier: Thermo Fisher Scientific
Danger

L04037.22 L04037.14
L04037.22EA 150 EUR
L04037.22 L04037.14
1,2-Bis(dimethylamino)methane 99%
1,2-Bis(dimethylamino)methane

Purity: 99% CAS No.: 51-80-9 Molecular Formula: C5H14N2 Formula Weight: 102.18 Flash Point: -12°(10°F) Refractive Index: 1.4010 Melting Point: -55° Boiling Point: 83-85° Beilstein Reference: 1731946 UN No.: 2733 MDL No.: MFCD00008328

Formula: C₅H₁₄N₂
Boiling Pt: 83…85 °C
Melting Pt: –55 °C
Density: 0,749
Flash Pt: –12 °C (10 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008328
CAS Number: 51-80-9
EINECS: 200-124-7
UN: 2733
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR