1-Chloro-2-phenylethane ≥99%

Supplier: Thermo Fisher Scientific
Warning

L04891.09 L04891.22 L04891.36
L04891.09EA 27 EUR
L04891.09 L04891.22 L04891.36
1-Chloro-2-phenylethane ≥99%
1-Chloro-2-phenylethane

Purity: 99% CAS No.: 622-24-2 Molecular Formula: C6H5(CH2)2Cl Formula Weight: 140.61 Flash Point: 66°(150°F) Refractive Index: 1.5300 Melting Point: -60° Boiling Point: 82-84°/16mm Beilstein Reference: 385969 MDL No.: MFCD00000977

Formula: ClCH₂CH₂C₆H₅
Boiling Pt: 82…84 °C (16 mmHg)
Melting Pt: –60 °C
Density: 1,070
Flash Pt: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000977
CAS Number: 622-24-2
EINECS: 210-725-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR