Diethyl-1-naphthylmethylphosphonate 90

Supplier: Thermo Fisher Scientific

Synonyms: Diethyl (naphthalen-1-ylmethyl)phosphonate

L05286.09 L05286.18
L05286.09EA 77.1 EUR
L05286.09 L05286.18
Diethyl-1-naphthylmethylphosphonate 90
Diethyl-1-naphthylmethylphosphonate

Purity: 95% CAS No.: 53575-08-9 Molecular Formula: C15H19O3P Formula Weight: 278.29 Refractive Index: 1.5650 Boiling Point: 205-206°/5mm Beilstein Reference: 2943656 MDL No.: MFCD00015769

Formula: C₁₅H₁₉O₃P
MW: 278.29 g/mol
Boiling Pt: 205…206 °C (5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00015769
CAS Number: 53575-08-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR