2,5-Diaminopyridine dihydrochloride 97%

Supplier: Thermo Fisher Scientific
Warning

L05599.06 L05599.14
L05599.06EA 55.4 EUR
L05599.06 L05599.14
2,5-Diaminopyridine dihydrochloride 97%
2,5-Diaminopyridine dihydrochloride

Purity: 97% CAS No.: 26878-35-3 Molecular Formula: C5H7N3.2HCl Formula Weight: 182.05 Melting Point: ca 264° dec. Beilstein Reference: 3912365

Formula: C₅H₉Cl₂N₃
Melting Pt: ∼ 264 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040818
CAS Number: 26878-35-3
EINECS: 248-083-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR