N-Ethylbenzylamine 97%

Supplier: Thermo Fisher Scientific
Danger

L05651.AC L05651.AE L05651.AP
L05651.ACEA 45.2 EUR
L05651.AC L05651.AE L05651.AP
N-Ethylbenzylamine 97%
N-Ethylbenzylamine

Purity: 97% CAS No.: 14321-27-8 Molecular Formula: C6H5CH2NHCH2CH3 Formula Weight: 135.21 Flash Point: 66°(150°F) Refractive Index: 1.5110 Boiling Point: 191-194° Beilstein Reference: 386023 UN No.: 2735 MDL No.: MFCD00009031

Formula: C₆H₅CH₂NHCH₂CH₃
Boiling Pt: 191…194 °C
Density: 0,910
Flash Pt: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009031
CAS Number: 14321-27-8
EINECS: 238-265-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR