Ethyl vanillate ≥97%

Supplier: Thermo Fisher Scientific

L05798.09 L05798.18
L05798.09EA 68.9 EUR
L05798.09 L05798.18
Ethyl vanillate ≥97%
Ethyl vanillate

Purity: 97% CAS No.: 617-05-0 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 42-46° Boiling Point: 293° MDL No.: MFCD00017269

Formula: C₁₀H₁₂O₄
Boiling Pt: 293 °C
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00017269
CAS Number: 617-05-0
EINECS: 210-503-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR