1,2:7,8-Diepoxyoctane ≥97%

Supplier: Thermo Fisher Scientific
Danger

L06768.09
L06768.09EA 98.1 EUR
L06768.09
1,2:7,8-Diepoxyoctane ≥97%
1,2:7,8-Diepoxyoctane

Purity: 97% CAS No.: 2426-07-5 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 97°(206°F) Refractive Index: 1.4450 Boiling Point: 238-240° Beilstein Reference: 104873 UN No.: 2922 MDL No.: MFCD00005155

Formula: C₈H₁₄O₂
Boiling Pt: 238…240 °C
Density: 0,997
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005155
CAS Number: 2426-07-5
EINECS: 219-375-9
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR