Diphenyl phosphoramidate ≥97%

Supplier: Thermo Fisher Scientific

L07155.06 L07155.14
L07155.06EA 70.4 EUR
L07155.06 L07155.14
Diphenyl phosphoramidate ≥97%
Diphenyl phosphoramidate

Purity: 97% CAS No.: 2015-56-7 Molecular Formula: C12H12NO3P Formula Weight: 249.21 Melting Point: 147-151° Beilstein Reference: 2054166

Formula: C₁₂H₁₂NO₃P
MW: 249.21 g/mol
MDL Number: MFCD00014078
CAS Number: 2015-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR