1-Octen-3-one 97% stabilised

Supplier: Thermo Fisher Scientific
Warning


Stabilisation: Stabilised with BHA 0,1%

L07189.03 L07189.06
L07189.03EA 65.1 EUR
L07189.03 L07189.06
1-Octen-3-one 97% stabilised
1-Octen-3-one

Stab. with 0.1% BHA


Purity: 97% CAS No.: 4312-99-6 Molecular Formula: C8H14O Formula Weight: 126.20 Storage Temperature: 2°C to 8°C Flash Point: 35°(95°F) Refractive Index: 1.4360 Odour: Characteristic Boiling Point: 56-60°/16mm Beilstein Reference: 1700905 UN No.: UN1224 MDL No.: MFCD00036558

Formula: C₈H₁₄O
Boiling Pt: 56…60 °C (16 mmHg)
Density: 0,848
Flash Pt: 35 °C (95 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00036558
CAS Number: 4312-99-6
EINECS: 224-327-5
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR