Methyl linoleate 95%

Supplier: Thermo Fisher Scientific

L07450.03 L07450.06
L07450.03EA 54 EUR
L07450.03 L07450.06
Methyl linoleate 95%
Methyl linoleate

Purity: 95% CAS No.: 112-63-0 Molecular Formula: C19H34O2 Formula Weight: 294.47 Storage Temperature: 2°C to 8°C Flash Point: 200°(392°F) Refractive Index: 1.4640 Melting Point: -35° Boiling Point: 207-208°/11mm Beilstein Reference: 1727614 Merck Reference: 14,6095 MDL No.: MFCD00009534

Formula: C₁₉H₃₄O₂
Boiling Pt: 207…208 °C (11 mmHg)
Melting Pt: –35 °C
Density: 0,888
Flash Pt: 200 °C (392 °F)
MDL Number: MFCD00009534
CAS Number: 112-63-0
EINECS: 203-993-0
Merck Index: 14,6095

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR