Ethidium bromide ≥98% (dry weight)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

L07482.03 L07482.06
L07482.03EA 46.9 EUR
L07482.03 L07482.06
Ethidium bromide ≥98% (dry weight)
Ethidium bromide

Purity: 98% CAS No.: 1239-45-8 Molecular Formula: C21H20BrN3 Formula Weight: 394.32 Odour: Weak Melting Point: ca 261° dec. Beilstein Reference: 3642536 Merck Reference: 14,4731 UN No.: 2811 MDL No.: MFCD00011724

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Boiling Pt: 72 °C (0.1 mmHg)
Melting Pt: ∼ 261 °C (dec.)
Density: 1.01 g/cm³ (20 °C)
Flash Pt: <100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6
UN: 2811
ADR: 6.1 I
Merck Index: 14,4731

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR