1,2-Epoxyhexane 96%

Supplier: Thermo Fisher Scientific
Danger

L07609.14 L07609.22
L07609.14EA 51.5 EUR
L07609.14 L07609.22
1,2-Epoxyhexane 96%
1,2-Epoxyhexane

Purity: 96% CAS No.: 1436-34-6 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 15°(59°F) Refractive Index: 1.4060 Boiling Point: 118-120° Beilstein Reference: 102568 UN No.: 1993 MDL No.: MFCD00005154

Formula: C₆H₁₂O
Boiling Pt: 118…120 °C
Density: 0,832
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005154
CAS Number: 1436-34-6
EINECS: 215-864-6
UN: 1993
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR