Lithium aluminium di-n-butylamide solution 0.16 M in 1,2-dimethoxyethane

Supplier: Thermo Fisher Scientific
Warning

L08269.AE L08269.AP
L08269.AEEA 145 EUR
L08269.AE L08269.AP
Lithium aluminium di-n-butylamide solution 0.16 M in 1,2-dimethoxyethane
Lithium aluminium di-n-butylamide

CAS No.: 15405-86-4 Molecular Formula: C32H72AlLiN4 Formula Weight: 546.87 Storage Temperature: 2°C to 8°C Melting Point: -54° UN No.: UN1993 MDL No.: MFCD00051918

Formula: C₃₂H₇₂AlLiN₄
Melting Pt: –54 °C
Density: 0,860
Storage Temperature: Refrigerator
MDL Number: MFCD00051918
CAS Number: 15405-86-4
UN: 1993
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR