N-(tert-Butoxycarbonyl)-L-aspartic acid 98%

Supplier: Thermo Fisher Scientific

Synonyms: N-Boc-L-aspartic Acid, Boc-Asp-OH

L08498.03 L08498.06
L08498.03EA 27.1 EUR
L08498.03 L08498.06
N-(tert-Butoxycarbonyl)-L-aspartic acid 98%
N-(tert-Butoxycarbonyl)-L-aspartic acid

Purity: 98% CAS No.: 13726-67-5 Molecular Formula: C9H15NO6 Formula Weight: 233.23 Storage Temperature: 2°C to 8°C Melting Point: 115° Beilstein Reference: 1913973 Optical Rotation: -5.5° (c=1 in methanol) MDL No.: MFCD00037279

Formula: C₉H₁₅NO₆
MW: 233.22 g/mol
Melting Pt: 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037279
CAS Number: 13726-67-5
EINECS: 237-294-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR