(4-Chlorophenylthio)acetic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

L08583.09 L08583.18
L08583.09EA 56.8 EUR
L08583.09 L08583.18
(4-Chlorophenylthio)acetic acid ≥98%
(4-Chlorophenylthio)acetic acid

Purity: 98% CAS No.: 3405-88-7 Molecular Formula: C8H7ClO2S Formula Weight: 202.66 Melting Point: 104-107° Beilstein Reference: 1871569

Formula: C₈H₇ClO₂S
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00021762
CAS Number: 3405-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR