N(α)-Boc-D-histidine ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Boc-D-His-OH

L08810.03 L08810.MD
L08810.03EA 87.6 EUR
L08810.03 L08810.MD
N(α)-Boc-D-histidine ≥98%
N(α)-Boc-D-histidine

Purity: 98+% CAS No.: 50654-94-9 Molecular Formula: C11H17N3O4 Formula Weight: 255.27 Storage Temperature: 2°C to 8°C Melting Point: ca 197° dec. Beilstein Reference: 4196575 Optical Rotation: -26° (c=1 in methanol) MDL No.: MFCD00037851

Formula: C₁₁H₁₇N₃O₄
MW: 255.3 g/mol
Melting Pt: ∼ 197 °C (dec.)
MDL Number: MFCD00037851
CAS Number: 50654-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR