Risocaine (propyl-4-aminobenzoate) 98%

Supplier: Thermo Fisher Scientific

Synonyms: Propyl 4-aminobenzoate

L09066.18 L09066.09 L09066.0B L09066.36
L09066.18EA 122 EUR
L09066.18 L09066.09 L09066.0B L09066.36
Risocaine (propyl-4-aminobenzoate) 98%
Risocaine (Propyl 4-aminobenzoate)

Purity: 98% CAS No.: 94-12-2 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 73-75° Beilstein Reference: 1211203

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00017111
CAS Number: 94-12-2
EINECS: 202-306-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR