Methyl-2-methylnicotinate ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: methyl 2-methylpyridine-3-carboxylate, Methyl 2-methylnicotinate

L09074.06 L09074.14
L09074.06EA 89.1 EUR
L09074.06 L09074.14
Methyl-2-methylnicotinate ≥97%
Methyl 2-methylnicotinate

Purity: 97% CAS No.: 65719-09-7 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Refractive Index: 1.5165 Boiling Point: 102-104°/10mm MDL No.: MFCD00051961

Formula: C₈H₉NO₂
Boiling Pt: 102…104 °C (10 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00051961
CAS Number: 65719-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR