Triethyl-2-phosphonobutyrate ≥97%

Supplier: Thermo Fisher Scientific
Warning

L09329.06 L09329.14
L09329.06EA 43.7 EUR
L09329.06 L09329.14
Triethyl-2-phosphonobutyrate ≥97%
Triethyl 2-phosphonobutyrate

Purity: 97% CAS No.: 17145-91-4 Molecular Formula: C10H21O5P Formula Weight: 252.25 Flash Point: 110°(230°F) Refractive Index: 1.4330 Boiling Point: 105-106°/2mm Beilstein Reference: 1789280 MDL No.: MFCD00041347

Formula: C₁₀H₂₁O₅P
Boiling Pt: 105…106 °C (2 mmHg)
Density: 1,064
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041347
CAS Number: 17145-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR