2-Butoxyethyl acetate ≥98%

Supplier: Thermo Fisher Scientific
Warning

L09515.AE L09515.AP L09515.0F
L09515.AEEA 26.9 EUR
L09515.AE L09515.AP L09515.0F
2-Butoxyethyl acetate ≥98%
2-Butoxyethyl acetate

Purity: 98% CAS No.: 112-07-2 Molecular Formula: C8H16O3 Formula Weight: 160.21 Flash Point: 71°(159°F) Refractive Index: 1.4130 Odour: Fruit-like Melting Point: -64° Boiling Point: 191-192° Beilstein Reference: 1756960 MDL No.: MFCD00009457

Formula: C₈H₁₆O₃
Boiling Pt: 191…192 °C
Melting Pt: –64 °C
Density: 0,942
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009457
CAS Number: 112-07-2
EINECS: 203-933-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR