(±)-1-Phenyl-2-propyn-1-ol 98%

Supplier: Thermo Fisher Scientific
Warning

L09549.03 L09549.06
L09549.03EA 35 EUR
L09549.03 L09549.06
(±)-1-Phenyl-2-propyn-1-ol 98%
(±)-1-Phenyl-2-propyn-1-ol

Purity: 98% CAS No.: 4187-87-5 Molecular Formula: C9H8O Formula Weight: 132.16 Flash Point: 99°(210°F) Refractive Index: 1.5500 Melting Point: 26-30° Boiling Point: 231-233° Beilstein Reference: 742365 MDL No.: MFCD00021860

Formula: C₉H₈O
Boiling Pt: 231…233 °C
Melting Pt: 26…30 °C
Density: 1,087
Flash Pt: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021860
CAS Number: 4187-87-5
EINECS: 224-064-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR