2,1,3-Benzothiadiazol-4-ylamine 98%

Supplier: Thermo Fisher Scientific
Danger

L09678.03 L09678.06
L09678.03EA 65.1 EUR
L09678.03 L09678.06
2,1,3-Benzothiadiazol-4-ylamine 98%
2,1,3-Benzothiadiazol-4-ylamine

Purity: 98% CAS No.: 767-64-6 Molecular Formula: C6H5N3S Formula Weight: 151.19 Melting Point: 67-69° Beilstein Reference: 3551

Formula: C₆H₅N₃S
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00005810
CAS Number: 767-64-6
EINECS: 212-186-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR