5-Isocyanato-1,2,3-trimethoxybenzene 97%

Supplier: Thermo Fisher Scientific
Danger

L09775.03 L09775.06
L09775.03EA 22.8 EUR
L09775.03 L09775.06
5-Isocyanato-1,2,3-trimethoxybenzene 97%
5-Isocyanato-1,2,3-trimethoxybenzene

Purity: 97% CAS No.: 1016-19-9 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Irritating Melting Point: 39-41° Boiling Point: 102-110°/0.4mm Beilstein Reference: 2736347 UN No.: UN2811 MDL No.: MFCD00013861

Formula: C₁₀H₁₁NO₄
Boiling Pt: 102…110 °C (0,4 mmHg)
Melting Pt: 39…41 °C
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00013861
CAS Number: 1016-19-9
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR