2-Chloro-5-(trifluoromethyl)phenyl isocyanate ≥97%

Supplier: Thermo Fisher Scientific
Danger

L09899.06 L09899.03
L09899.06EA 72.7 EUR
L09899.06 L09899.03
2-Chloro-5-(trifluoromethyl)phenyl isocyanate ≥97%
2-Chloro-5-(trifluoromethyl)phenyl isocyanate

Purity: 97% CAS No.: 50528-86-4 Molecular Formula: C8H3ClF3NO Formula Weight: 221.57 Storage Temperature: 2°C to 8°C Flash Point: 92°(197°F) Refractive Index: 1.4890 Boiling Point: 52-53°/3mm Beilstein Reference: 2112804 UN No.: 2206 MDL No.: MFCD00037029

Formula: C₈H₃ClF₃NO
Boiling Pt: 52…53 °C (3 mmHg)
Density: 1,480
Flash Pt: 92 °C (197 °F)
MDL Number: MFCD00037029
CAS Number: 50528-86-4
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR