Tetrahydrofurfuryl acrylate 90% stabilised, tech.

Supplier: Thermo Fisher Scientific
Warning


Stabilisation: Stabilised with hydroquinone (HQ) 500 ppm

L10299.18 L10299.30
L10299.18EA 39.3 EUR
L10299.18 L10299.30
Tetrahydrofurfuryl acrylate 90% stabilised, tech.
Tetrahydrofurfuryl acrylate

Stab. with 500ppm hydroquinone


Purity: 90% CAS No.: 2399-48-6 Molecular Formula: C8H12O3 Formula Weight: 156.18 Flash Point: 93°(199°F) Refractive Index: 1.4550 Boiling Point: 88-89°/9mm Beilstein Reference: 113115 UN No.: UN3334 MDL No.: MFCD00014516

Formula: C₈H₁₂O₃
Boiling Pt: 88…89 °C (9 mmHg)
Density: 1,030
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014516
CAS Number: 2399-48-6
EINECS: 219-268-7
UN: 3334
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR