1,2-Benzenedithiol ≥97%

Supplier: Thermo Fisher Scientific
Danger

L11253.03 L11253.06 L11253.MD
L11253.03EA 146 EUR
L11253.03 L11253.06 L11253.MD
1,2-Benzenedithiol ≥97%
1,2-Benzenedithiol

Purity: 97% CAS No.: 17534-15-5 Molecular Formula: C6H6S2 Formula Weight: 142.24 Storage Temperature: 2°C to 8°C Flash Point: 103°(217°F) Refractive Index: 1.6580 Odour: Unpleasant Melting Point: 21-25° Boiling Point: 239° Beilstein Reference: 636154 UN No.: 3335 MDL No.: MFCD00004835

Formula: C₆H₆S₂
Boiling Pt: 239 °C
Melting Pt: 21…25 °C
Density: 1,236
Flash Pt: 103 °C (217 °F)
MDL Number: MFCD00004835
CAS Number: 17534-15-5
EINECS: 241-530-4
ADR: 9,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR