3,5-Dimethylphenyl isocyanate ≥97%

Supplier: Thermo Fisher Scientific
Danger

L11698.03 L11698.06
L11698.03EA 31.8 EUR
L11698.03 L11698.06
3,5-Dimethylphenyl isocyanate ≥97%
3,5-Dimethylphenyl isocyanate

Purity: 97% CAS No.: 54132-75-1 Molecular Formula: C9H9NO Formula Weight: 147.18 Storage Temperature: 2°C to 8°C Flash Point: 83°(181°F) Refractive Index: 1.5280 Boiling Point: 205° Beilstein Reference: 774958 UN No.: 2206 MDL No.: MFCD00013868

Formula: C₉H₉NO
Boiling Pt: 205 °C
Density: 1,045
Flash Pt: 83 °C (181 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013868
CAS Number: 54132-75-1
EINECS: 258-987-0
UN: 2206
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR