5-Hydroxy-2-nitrobenzaldehyde ≥97%

Supplier: Thermo Fisher Scientific
Warning

L12051.03 L12051.06
L12051.03EA 0 EUR
L12051.03 L12051.06
5-Hydroxy-2-nitrobenzaldehyde ≥97%
5-Hydroxy-2-nitrobenzaldehyde

Purity: 97% CAS No.: 42454-06-8 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 166-168° Beilstein Reference: 1910196

Formula: C₇H₅NO₄
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00007332
CAS Number: 42454-06-8
EINECS: 255-832-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR